missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Talc, Powder, Multi-Compendial, J.T. Baker™
Click to view available options
Quantity:
500 g
2 kg
Packaging:
Poly Bottle
Specifications
Specifications
Packaging | Poly Bottle |
Quantity | 2 kg |
Grade | Multi-Compendial |
Physical Form | Powder |
Chemical Name or Material | Talc |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction