missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TADA1L Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 € - 410.00 €
Specifications
Antigen | TADA1L |
---|---|
Concentration | 0.59 mg/mL |
Applications | Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16819864
|
Proteintech
20337-1-AP-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16809864
|
Proteintech
20337-1-AP-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The MW of this protein is 37 kDa, and 20337-1-AP specifically recognizes the 37 kDa protein.
Probably involved in transcriptional regulation.Specifications
TADA1L | |
Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Mouse, Human | |
Q96BN2, Q99LM9 | |
117143, 27878 | |
TADA1L Fusion Protein Ag14118 | |
Primary | |
-20°C | |
TADA1 |
0.59 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
KIAA0764, RP1 9E21.4, SPT3 associated factor 42, STAF42, TADA1, TADA1L, Transcriptional adapter 1 | |
Tada1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title