missing translation for 'onlineSavingsMsg'
Learn More

IKA™ S 10 N Dispersing Tool - ST

Suitable for dispersing instrument T 10 basic and T 10 standard

Brand:  IKA™ 0004446500

 View more versions of this product

Product Code. 13460597

  • 1575.00 € / Each
Estimated Shipment: 19-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

ST tools are recommended for use with tissue and other fibrous materials
Specifications

Specifications

Dispersing Element
0.3 in.
115 mm
S 10 N 8 G ST
1 Ea.
1 to 50 mL
8 mm
PTFE, 316L Stainless Steel PT
S 10N-8G-ST
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers