Learn More
Sigma Laborzentrifugen™ Swing-out Rotors for Sigma™ Series 3 Centrifuges
Use these rotors with maximum load and at maximum speed for up to 50,000 cycles with low maintenance. Sigma™ Swing-out Rotors for Sigma™ Series 3 Centrifuges are ideal for use in medical and research laboratories.
Description
- Swing-bucket rotors for sedimentation with smaller centrifugal forces of up to approximately 6000 x g
- A higher yield may be obtained as it is possible to fill the centrifuge test tube completely
- Sedimentation takes place in the middle of the bottom of the test tube enabling easy separation of the phases
- Sigma supplies matching fixed-angle, swing-out and drum rotors for all Sigma centrifuges
- Sigma rotors are precision-made from modern, certified high-performance materials with traceable batch numbers
- Thanks to many years of experience and close contact with our customers, we are able to offer an extensive selection of rotors
- Sigma rotors are characterized by extra capacity or higher maximum speed
- Rotors can be operated continuously at maximum speed and +4°C in nearly all Sigma centrifuges
Specifications
Swing-out Rotor |