Learn More
SVL™ Cone/Screw Thread Adapter
Facilitates connection between SVL™ and Quickfit™ glassware.
Brand:  SVL 670-77
 View more versions of this product
Product Code. 15587740
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Features an SVL™ screwthread and Quickfit™ ground glass socket
  • Thread accepts SVL™ screw caps and couplings. Cone fits Quickfit™ sockets
  • Manufactured from temperature and chemical resistant borosilicate glass
TRUSTED_SUSTAINABILITY
Specifications
Specifications
24/29
Borosilicate Glass
22
SVL™ and Quickfit™ Glassware
Cone/Screw thread adapter