Learn More
Sampling Systems™ Surface Catch Net
Make removing floating objects during water sampling quick and easy with this surface catch net attachment for the TeleScoop.
Brand:  Sampling Systems™ 5065M-310
Product Code. 17982042
  • 64.90 € / Each
Explore more special offers
Description
Description
  • Telescopic samplers are often used for taking surface samples from rivers, tanks and lakes
  • A wide choice of attachments is available to make the TeleScoop ideally suited for most applications
  • Attachments are interchangeable and simply click into position
Specifications
Specifications
Swabbing Template
Not Autoclavable
No
Polypropylene
Reusable
Non-sterile