Learn More
Berkshire Super Polx™ SWSE
BIOPHARMA
Super Polx™ SWSE is a sealed edge cleanroom laundered wiper recommended for ISO Class 3 and above environments.
Brand:  Berkshire SPSWSE121210
 View more versions of this product
Product Code. 15540682
  • 1280.00 € / Pack of 1500
Explore more special offers
Description
Description
  • 100% continuous filament double knitted polyester
  • Ultrasonically sealed edge
  • Laundered and packaged in an ISO Class 4 (Class 10) cleanroom
  • Lot tracing on each pack
  • Economical
  • Strong, tear and abrasion resistant
  • Very good absorbency
  • Low particles and fibers
  • Soft hand and edge for sensitive surfaces
Specifications
Specifications
Surface Wiper
30 cm
30 cm
Not Autoclavable
1-ply
11.8 in.
12 in.
Polyester
Single-use
Non-sterile