missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid Solution 2M (4N), Pure, Bench Reagent, Fisher Chemical™

Product Code. 11478962
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Bag in box
HDPE plastic bottle
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

H2O4S, CAS Number-7664-93-9, acide sulfurique, battery acid, vitriol brown oil, electrolyte acid, dipping acid, acidum sulfuricum, sulphuric acid, mattling acid, dihydrogen sulfate, oil of vitriol, 1L, 120 deg.C, CHEBI:26836, Colorless, QAOWNCQODCNURD-UHFFFAOYSA-N, sulfuric acid, 64589, 98.072

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

pH 1
Color Colorless
Boiling Point 120°C
Packaging Bag in box
Quantity 10 L
Physical Form Liquid
Chemical Name or Material Sulfuric Acid Solution
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'