missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric acid solution, Volumetric, Reag. Ph. Eur., 0.5 M H2SO4 (1.0N), Honeywell Fluka™

Volumetric, Reag. Ph. Eur., 0.5 M H2SO4 (1.0N)

39.02 € - 343.85 €

Chemical Identifiers

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O
View More Specs

Products 8
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15634980
View Documents
Honeywell-Fluka
35354-1L
1 L Plastic Bottle
39.02 €
1 L
Please to purchase this item. Need a web account? Register with us today!
15644980
View Documents
Honeywell-Fluka
35354-2.5L
2.5 L Plastic Bottle
81.55 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
15674980
View Documents
Honeywell-Fluka
35354-5L
5 L Plastic Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15694980
View Documents
Honeywell-Fluka
35354-6X1L
6 x 1 L Plastic Bottle
197.19 €
Pack of 6
Please to purchase this item. Need a web account? Register with us today!
15624980
View Documents
Honeywell-Fluka
35354-10L-VP
10 L VOLPAC™ CONTAINER
198.98 €
10 L
Please to purchase this item. Need a web account? Register with us today!
15654980
View Documents
Honeywell-Fluka
35354-4X2.5L
4 x 2.5 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15614980
View Documents
Honeywell-Fluka
35354-10L
10 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15664980
View Documents
Honeywell-Fluka
35354-4X5L
4 x 5 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

Specifications

Chemical Identifiers

7664-93-9
98.07
QAOWNCQODCNURD-UHFFFAOYSA-N
CHEBI:26836
OS(O)(=O)=O
H2O4S
MFCD00064589
1118
sulfuric acid

Specifications

7664-93-9
H2O4S
MFCD00064589
2037554
QAOWNCQODCNURD-UHFFFAOYSA-N
sulfuric acid
1118
98.08g/mol
Sulfuric acid solution
0.5M
H2SO4
UN3264
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
OS(O)(=O)=O
98.07
CHEBI:26836
Reagent Ph. Eur.
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Sulfuric acid solution

Signal Word
  • Warning

Hazard Category
  • Substances and mixtures corrosive to metals Category 1

Hazard Statement
  • H290-May be corrosive to metals.

Precautionary Statement
  • P234-Keep only in original container.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

Supplemental information
  • MIXTURE LIST-Contains : sulphuric acid

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.