missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric Acid Solution 0.05M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™
Click to view available options
Quantity:
1 L
2.5 L
10 L
25 L
Packaging:
Bag in box
HDPE plastic bottle
Plastic container
Specifications
Specifications
Color | Colorless |
Boiling Point | 100°C |
Packaging | Plastic container |
Vapor Pressure | 1mmHg at 20°C |
Quantity | 25 L |
Physical Form | Liquid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'