missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric Acid, Pure, d=1.25, for Batteries And Accumulators, Fisher Chemical
Click to view available options
Quantity:
2.5 L
Packaging:
HDPE plastic bottle
Specifications
Specifications
pH | 1 |
Color | Colorless |
CAS | 7664-93-9 |
Boiling Point | 120°C |
Packaging | HDPE plastic bottle |
Quantity | 2.5 L |
Molecular Formula | H2O4S |
Physical Form | Liquid |
MDL Number | MFCD00064589 |
Synonym | oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil |
Show More |
Safety and Handling

Signal Word
- Danger
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- P234-Keep only in original container.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : Sulfuric acid
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction