missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric Acid, Pure, 25% (v/v), Fisher Chemical
Brand: Fisher Chemical H/0564/21
This item is not returnable.
View return policy
Description
5L, HDPE plastic bottle
Specifications
1 | |
7664-93-9 | |
HDPE plastic bottle | |
H2O4S | |
MFCD00064589 | |
QAOWNCQODCNURD-UHFFFAOYSA-N | |
sulfuric acid | |
1118 |
Colorless | |
120°C | |
5 L | |
Liquid | |
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
OS(O)(=O)=O | |
98.07 | |
CHEBI:26836 |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Sulfuric acid, 25% w/v
Signal Word
- Danger
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P234-Keep only in original container.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : Sulfuric acid