Learn More
Sulfuric acid, for analysis ACS, 95% solution in water, Thermo Scientific Chemicals
30.75 € - 194.00 €
Chemical Identifiers
CAS | 7664-93-9 |
---|---|
Molecular Formula | H2O4S |
Molecular Weight (g/mol) | 98.07 |
MDL Number | MFCD00064589 |
InChI Key | QAOWNCQODCNURD-UHFFFAOYSA-N |
Synonym | oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil |
PubChem CID | 1118 |
ChEBI | CHEBI:26836 |
IUPAC Name | sulfuric acid |
SMILES | OS(O)(=O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10351741
|
Thermo Scientific Acros
424525000 |
500 mL | Glass Bottle |
30.75 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10391313
|
Thermo Scientific Acros
424525001 |
500 mL | Plastic bottle |
31.36 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10015910
|
Thermo Scientific Acros
424520025 |
2.5 L | HDPE Bottles |
93.34 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10536933
|
Thermo Scientific Acros
424520026 |
2.5 L | Plastic bottle |
79.90 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11914091
|
Thermo Scientific Acros
424520100 |
10 L | Plastic drum |
194.00 €
10 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7664-93-9 | |
98.07 | |
QAOWNCQODCNURD-UHFFFAOYSA-N | |
1118 | |
sulfuric acid |
H2O4S | |
MFCD00064589 | |
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
CHEBI:26836 | |
OS(O)(=O)=O |
Specifications
7664-93-9 | |
290.0°C | |
1ppm max. | |
1.83 | |
H2O4S | |
H2SO4 | |
MFCD00064589 | |
15, 9104 | |
Solubility in water: miscible. Other solubilities: miscible with alcohol (generation of heat) | |
OS(O)(=O)=O | |
98.07 | |
CHEBI:26836 | |
95.0 to 98.0% | |
Sulfuric acid |
10.0°C | |
1.8300g/mL | |
0.0005% max. | |
95% | |
21 mPa.s (25°C) | |
Liquid Free From Suspended or Insoluble Matter | |
01,470; 05,633; 06,558; 07,347; 09,441 | |
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
QAOWNCQODCNURD-UHFFFAOYSA-N | |
sulfuric acid | |
1118 | |
98.08 | |
ACS Reagent |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.