Learn More
Sulfuric acid, 99.9999% (metals basis), 92% min, Thermo Scientific Chemicals
52.40 € - 814.00 €
Chemical Identifiers
CAS | 7664-93-9 |
---|---|
Molecular Formula | H2O4S |
Molecular Weight (g/mol) | 98.07 |
MDL Number | MFCD00064589 |
InChI Key | QAOWNCQODCNURD-UHFFFAOYSA-N |
Synonym | oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil |
PubChem CID | 1118 |
ChEBI | CHEBI:26836 |
IUPAC Name | sulfuric acid |
SMILES | OS(O)(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11341608
|
Thermo Scientific Alfa Aesar
011000.AB |
10 mL |
52.40 €
10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11351608
|
Thermo Scientific Alfa Aesar
011000.AE |
100 mL |
223.00 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11345819
|
Thermo Scientific Alfa Aesar
011000.AP |
500 mL |
814.00 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sulfuric acid is used as an electrolyte in lead-acid batteries, and a cleaning agent. It is used in mineral processing, fertilizer manufacturing, oil refining, wastewater processing, and chemical synthesis. It is also used in the production of detergents, synthetic resins, dyestuffs, pharmaceuticals, petroleum catalysts, insecticides, antifreeze, processes such as oil well acidizing, aluminum reduction, paper sizing, pigments, enamels, printing inks, coated fabrics and paper. It is also used in the production of explosives, cellophane, acetate and viscose textiles, lubricants, non-ferrous metals and batteries. Sulfuric acid immobilized on silica gel is used as an efficient and benign catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7664-93-9 | |
98.07 | |
QAOWNCQODCNURD-UHFFFAOYSA-N | |
1118 | |
sulfuric acid |
H2O4S | |
MFCD00064589 | |
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
CHEBI:26836 | |
OS(O)(=O)=O |
Specifications
Colorless to Yellow | |
≤92% | |
Acrid | |
Liquid | |
UN1830 | |
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
QAOWNCQODCNURD-UHFFFAOYSA-N | |
sulfuric acid | |
1118 | |
98.08 | |
(metals basis) |
7664-93-9 | |
1.84 g/mL | |
H2O4S | |
MFCD00064589 | |
14,8974 | |
Sulfuric acid is miscible with water at all concentrations. | |
OS(O)(=O)=O | |
98.07 | |
CHEBI:26836 | |
99.9999% | |
Sulfuric acid |
Safety and Handling
GHS H Statement
H314
Causes severe skin burns and eye damage.
P201-P202-P260-P264b-P271-P280-P285-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P310-P363-P501c
H314-H330-H350
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: SULFURIC ACID
missing translation for 'einecsNumber' : 231-639-5
missing translation for 'rtecsNumber' : WS5600000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only