missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric acid 62%, d=1.52, for the determination of fats in cheese, Fisher Chemical™
Suitable for the determination of fats in cheese.
51.70 € - 185.00 €
Chemical Identifiers
CAS | 7664-93-9, 7732-18-5 |
---|---|
Molecular Formula | H2O4S |
Molecular Weight (g/mol) | 98.07 |
MDL Number | MFCD00064589 |
InChI Key | QAOWNCQODCNURD-UHFFFAOYSA-N |
IUPAC Name | sulfuric acid |
SMILES | OS(O)(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15948916
|
Fisher Chemical
S/9370/15 |
1 L |
51.70 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15958916
|
Fisher Chemical
S/9370/21 |
5 L |
185.00 €
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
7664-93-9, 7732-18-5 | |
98.07 | |
QAOWNCQODCNURD-UHFFFAOYSA-N | |
OS(O)(=O)=O |
H2O4S | |
MFCD00064589 | |
sulfuric acid |
Specifications
7664-93-9, 7732-18-5 | |
HDPE Plastic Bottle | |
H2O4S | |
MFCD00064589 | |
OS(O)(=O)=O | |
98.07 |
1.52 | |
61.5-62.5% | |
Liquid | |
QAOWNCQODCNURD-UHFFFAOYSA-N | |
sulfuric acid |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Sulphuric acid 62% for the determination of fats in cheese (Gerber test)
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : sulphuric acid
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title