missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfur pieces, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
52.20 € - 626.00 €
Chemical Identifiers
CAS | 7704-34-9 |
---|---|
Molecular Formula | S |
Molecular Weight (g/mol) | 32.06 |
MDL Number | MFCD00085316 |
InChI Key | NINIDFKCEFEMDL-UHFFFAOYSA-N |
Synonym | polymeric, lac, ad usum externum, acmc-20a47y, rubbervulcanizationagentis ot |
PubChem CID | 5362487 |
ChEBI | CHEBI:26833 |
IUPAC Name | sulfur |
SMILES | [S] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11346847
|
Thermo Scientific Alfa Aesar
010755.09 |
10 g |
52.20 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11325809
|
Thermo Scientific Alfa Aesar
010755.18 |
50 g |
146.00 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11356847
|
Thermo Scientific Alfa Aesar
010755.30 |
250 g |
626.00 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7704-34-9 | |
32.06 | |
NINIDFKCEFEMDL-UHFFFAOYSA-N | |
5362487 | |
sulfur |
S | |
MFCD00085316 | |
polymeric, lac, ad usum externum, acmc-20a47y, rubbervulcanizationagentis ot | |
CHEBI:26833 | |
[S] |
Specifications
7704-34-9 | |
Pieces | |
S | |
polymeric, lac, ad usum externum, acmc-20a47y, rubbervulcanizationagentis ot | |
[S] | |
32.06 | |
CHEBI:26833 | |
Pale yellow pieces | |
Sulphur |
Yellow | |
99.999% (Metals basis) | |
MFCD00085316 | |
NINIDFKCEFEMDL-UHFFFAOYSA-N | |
sulfur | |
5362487 | |
Weak | |
2.07g/cm3 |
Safety and Handling
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319-H500
missing translation for 'dotInformation' : DOT Class: 4.1, Packaging Group: III
missing translation for 'einecsNumber' : 231-722-6
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only