missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfur Oil Standard in No. 2 Diesel Fuel, 10μg/g, Bulk, SPEX CertiPrep™
Shop All SPEX Certiprep Products
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mL
Packaging:
Triple-leached LDPE bottle
Description
Includes
NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
- Sulfur Oil Standard Blank
- For use in diesel fuel analysis
- 1 mL volume in #2 Diesel Fuel
Certifications
ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015
Compliance
GHS/OSHA Complaint
Specifications
Specifications
CAS | 68476-34-6 |
Physical Form | Liquid |
Quantity | 100 mL |
Packaging | Triple-leached LDPE bottle |
Chemical Name or Material | Sulfur Oil Standard in #2 Diesel Fuel, For Diesel Fuel Analysis |
Safety and Handling


Product Identifier
- Sulfur Oil Standard in 2 Diesel Fuel
- Warning
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- H226-Flammable liquid and vapour.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H351-Suspected of causing cancer.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Fuels, diesel, No 2
Product intended for laboratory use only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sulfur Oil Standard in No. 2 Diesel Fuel, 10μg/g, Bulk, SPEX CertiPrep™ >
Spot an opportunity for improvement?Share a Content Correction