missing translation for 'onlineSavingsMsg'
Learn More

Sulfur in Isooctane standard solution, Specpure™, 5μg/g (0.0005%), Thermo Scientific Chemicals

Product Code. 15426895
Click to view available options
Quantity:
100 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 100 mL
MDL Number MFCD00804269
UN Number UN1262
Grade Specpure™
Odor Petrol-like
Chemical Name or Material Sulfur in Isooctane standard solution, 5μg/g (0.0005%)

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'