missing translation for 'onlineSavingsMsg'
Learn More

Sulfapyridine, 95%, Thermo Scientific Chemicals

47.84 € - 153.77 €

Chemical Identifiers

CAS 144-83-2
Molecular Formula C11H11N3O2S
Molecular Weight (g/mol) 249.288
InChI Key GECHUMIMRBOMGK-UHFFFAOYSA-N
PubChem CID 5336
ChEBI CHEBI:132842
IUPAC Name 4-amino-N-pyridin-2-ylbenzenesulfonamide
SMILES C1=CC=NC(=C1)NS(=O)(=O)C2=CC=C(C=C2)N
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15422152
SDS View Documents
Thermo Scientific Acros
442850250
25 g
47.84 €
25g
Please to purchase this item. Need a web account? Register with us today!
15432152
SDS View Documents
Thermo Scientific Acros
442851000
100 g
153.77 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

144-83-2
249.288
5336
4-amino-N-pyridin-2-ylbenzenesulfonamide
C11H11N3O2S
GECHUMIMRBOMGK-UHFFFAOYSA-N
CHEBI:132842
C1=CC=NC(=C1)NS(=O)(=O)C2=CC=C(C=C2)N

Specifications

Sulfapyridine
190.0°C
0.5% max.
GECHUMIMRBOMGK-UHFFFAOYSA-N
4-amino-N-pyridin-2-ylbenzenesulfonamide
5336
249.29
Sulfapyridine
144-83-2
Authentic
C11H11N3O2S
C1=CC=NC(=C1)NS(=O)(=O)C2=CC=C(C=C2)N
249.288
CHEBI:132842
95%
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Sulfapyridine

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sulfapyridine, 95%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.