missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUGT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11675-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene is homologous to the yeast gene SGT1, which encodes a protein involved in kinetochore function and required for the G1/S and G2/M transitions. Complementation studies suggest that the human protein has similar functions. Two transcript variants encoding different isoforms have been found for this gene.Specifications
SUGT1 | |
Polyclonal | |
Unconjugated | |
SUGT1 | |
Protein 40 6 3, SGT1, SUGT1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10910, 290408, 67955 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
B0BN85, Q9CX34, Q9Y2Z0 | |
Sugt1 | |
SUGT1 Fusion Protein Ag2275 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction