missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sudan Red 7B

56.78 € - 316.58 €

Chemical Identifiers

CAS 6368-72-5
Molecular Formula C24H21N5
Molecular Weight (g/mol) 379.467
MDL Number MFCD00003904
InChI Key VKWNTWQXVLKCSG-UHFFFAOYSA-N
Synonym C.I. 26050, N-Ethyl-1-[4-(phenylazo)phenylazo]-2-naphthylamine, Fat Red 7B, Solvent Red 19
PubChem CID 61396
IUPAC Name N-ethyl-1-[(4-phenyldiazenylphenyl)diazenyl]naphthalen-2-amine
SMILES CCNC1=C(C2=CC=CC=C2C=C1)N=NC3=CC=C(C=C3)N=NC4=CC=CC=C4
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10366691
SDS View Documents
Thermo Scientific Chemicals
191630050
5 g Glass bottle
56.78 €
5g
Please to purchase this item. Need a web account? Register with us today!
10541831
SDS View Documents
Thermo Scientific Chemicals
191630500
50 g Glass bottle
316.58 €
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Sudan Red 7B

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.