missing translation for 'onlineSavingsMsg'
Learn More

Sudan orange G, pure, Thermo Scientific Chemicals

Product Code. 10481582
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2051-85-6
Molecular Formula C12H10N2O2
Molecular Weight (g/mol) 214.22
MDL Number MFCD00002275
InChI Key PULNESMFDLBKAZ-SDNWHVSQSA-N
Synonym C.I. 11920, 4-(Phenylazo)resorcinol, Solvent orange 1
PubChem CID 5921655
IUPAC Name (4Z)-3-hydroxy-4-(phenylhydrazinylidene)cyclohexa-2,5-dien-1-one
SMILES OC1=CC(=O)C=C\C1=N/NC1=CC=CC=C1

Specifications

Infrared Spectrum Authentic
Formula Weight 214.22
Grade Extra Pure
Packaging Glass bottle
UV Spectrometry Complies
Color Orange to Red
Quantity 25 g
Physical Form Powder
Chemical Name or Material Sudan orange G, Pure
compliance-icons
Product Identifier
  • Sudan Orange G
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.