missing translation for 'onlineSavingsMsg'
Learn More

Techne™ External Bypass for Circulators

Designed for use when temperatures down to -35°C are required

Brand:  Techne™ 36620-88

Product Code. 11573759

  • 407.00 € / Each
Estimated Shipment: 23-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Compact refrigeration units with a 200 mL internal capacity
  • Designed for use with Techne liquid baths
Specifications

Specifications

C-85/FC-200/FC-500
Bypass
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers