Learn More
Strontium peroxide, 12.3% available oxygen, Thermo Scientific Chemicals
79.29 € - 1077.36 €
Chemical Identifiers
CAS | 1314-18-7 |
---|---|
Molecular Formula | O2Sr |
Molecular Weight (g/mol) | 119.618 |
MDL Number | MFCD00084686 |
InChI Key | UHCGLDSRFKGERO-UHFFFAOYSA-N |
Synonym | strontium peroxide, 1,2-dioxa-3-strontacyclopropane, strontium dioxide, strontiumperoxid, strontium peroxide sr o2, hsdb 788, strontium peroxide un1509 oxidizer, strontium peroxide available oxygenShow More |
PubChem CID | 14807 |
IUPAC Name | strontium;peroxide |
SMILES | [O-][O-].[Sr+2] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15442647
|
Thermo Scientific Alfa Aesar
046310.30 |
250 g |
79.29 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15452647
|
Thermo Scientific Alfa Aesar
046310.A1 |
1 kg |
251.25 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15462647
|
Thermo Scientific Alfa Aesar
046310.A7 |
5 kg |
1077.36 €
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Applications of Strontium Peroxide include pyrotechnics i.e. fireworks (red color), ammunitions (tracer/hunting), oxidizer, alarm rocket. It is also used in textiles for fibers bleaching and as an oxidizing agent for hair bleaching (professional).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1314-18-7 | |
119.618 | |
UHCGLDSRFKGERO-UHFFFAOYSA-N | |
14807 | |
[O-][O-].[Sr+2] |
O2Sr | |
MFCD00084686 | |
strontium peroxide, 1,2-dioxa-3-strontacyclopropane, strontium dioxide, strontiumperoxid, strontium peroxide sr o2, hsdb 788, strontium peroxide un1509 oxidizer, strontium peroxide available oxygenShow More | |
strontium;peroxide |
Specifications
215°C (decomposition) | |
Brown | |
O2Sr | |
MFCD00084686 | |
14,8847 | |
Insoluble in water and slowly decomposes in water. | |
[O-][O-].[Sr+2] | |
119.618 | |
119.62 | |
Strontium peroxide, 12.3% available oxygen |
1314-18-7 | |
Powder | |
SrO2 | |
UN1509 | |
strontium peroxide, 1,2-dioxa-3-strontacyclopropane, strontium dioxide, strontiumperoxid, strontium peroxide sr o2, hsdb 788, strontium peroxide un1509 oxidizer, strontium peroxide available oxygen | |
UHCGLDSRFKGERO-UHFFFAOYSA-N | |
strontium;peroxide | |
14807 | |
Moisture sensitive |
Safety and Handling
GHS H Statement
H272-H318-H335-H315
May intensify fire
oxidizer.
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'dotInformation' : Hazard Class: 5.1
missing translation for 'einecsNumber' : 215-224-6
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Strontium peroxide
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash face, hands, and anyexposed skin thoroughly after handling.
- P271-Use only outdoors or in a well-ventilated area.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Strontium peroxide
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.