missing translation for 'onlineSavingsMsg'
Learn More

Strontium nitrate, anhydrous, 98%, Thermo Scientific Chemicals

45.83 € - 211.05 €

Chemical Identifiers

CAS 10042-76-9
Molecular Formula N2O6Sr
Molecular Weight (g/mol) 211.63
MDL Number MFCD00011248
InChI Key DHEQXMRUPNDRPG-UHFFFAOYSA-N
Synonym strontium nitrate, strontium dinitrate, nitric acid, strontium salt, strontium ii nitrate 1:2, unii-bdg873aqzl, nitrate de strontium french, hsdb 787, strontium nitrate sr no3 2, bdg873aqzl, nitric acid, strontium salt 2:1
PubChem CID 24848
IUPAC Name strontium;dinitrate
SMILES [Sr++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11397298
View Documents
Thermo Scientific Alfa Aesar
030534.30
250 g
45.83 €
250g
Please to purchase this item. Need a web account? Register with us today!
11387298
View Documents
Thermo Scientific Alfa Aesar
030534.A1
1 kg
73.06 €
1kg
Please to purchase this item. Need a web account? Register with us today!
11307308
View Documents
Thermo Scientific Alfa Aesar
030534.D9
5 x 1 kg
211.05 €
5kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

In railroad flares, signal lights, marine signalsStrontium nitrate is used in the production of road flares and in fireworks. It serves as a colorant (red) in pyrotechnics and is also used as an oxidizer in pyrotechnics. Mixed with glycolic acid, it is used to reduce the sensation of skin irritation. It is also used in electrophysiology experiments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10042-76-9
211.63
DHEQXMRUPNDRPG-UHFFFAOYSA-N
24848
[Sr++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
N2O6Sr
MFCD00011248
strontium nitrate, strontium dinitrate, nitric acid, strontium salt, strontium ii nitrate 1:2, unii-bdg873aqzl, nitrate de strontium french, hsdb 787, strontium nitrate sr no3 2, bdg873aqzl, nitric acid, strontium salt 2:1
strontium;dinitrate

Specifications

570°C
White
98%
MFCD00011248
Freely soluble in water. Slightly soluble in alcohol and acetone.Soluble in water and ammonia. Slightly soluble in alcohol,ethanol and acetone. Insoluble in nitric acid.
[Sr++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
211.63
211.63
Hygroscopic
Strontium nitrate, Anhydrous
10042-76-9
Granules
N2O6Sr
strontium nitrate, strontium dinitrate, nitric acid, strontium salt, strontium ii nitrate 1:2, unii-bdg873aqzl, nitrate de strontium french, hsdb 787, strontium nitrate sr no3 2, bdg873aqzl, nitric acid, strontium salt 2:1
DHEQXMRUPNDRPG-UHFFFAOYSA-N
strontium;dinitrate
24848
Odorless
2.99
Videos
Safety and Handling

Safety and Handling

P210-P220-P221-P280-P305+P351+P338-P310-P370+P378q-P501c

H272-H318

missing translation for 'dotInformation' : Hazard Class: 5.1; Packaging Group: III

missing translation for 'einecsNumber' : 233-131-9

missing translation for 'rtecsNumber' : WK9800000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Strontium nitrate

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Oxidising solid Category 1

Hazard Statement
  • H271-May cause fire or explosion; strong oxidiser.
  • H318-Causes serious eye damage.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P370+P380+P375-In case of fire: Evacuate area. Fight fire remotely due to the risk of explosion.

Supplemental information
  • MIXTURE LIST-Contains : Strontium nitrate

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.