Learn More
Strontium isopropoxide, 96%, Thermo Scientific Chemicals
62.41 € - 536.67 €
Chemical Identifiers
CAS | 88863-33-6 |
---|---|
Molecular Formula | C6H14O2Sr |
Molecular Weight (g/mol) | 205.80 |
MDL Number | MFCD00050487 |
InChI Key | OHULXNKDWPTSBI-UHFFFAOYSA-N |
Synonym | strontium isopropoxide, strontium 2+ bis propan-2-olate, strontium propan-2-olate, diisopropoxystrontium, acmc-20egm0, strontium isopropylate, 2-propanol, strontiumsalt 9ci, strontium 2+ ion bis propan-2-olate |
PubChem CID | 6099237 |
SMILES | [Sr++].CC(C)[O-].CC(C)[O-] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11372428
|
Thermo Scientific Alfa Aesar
014481.03 |
1 g |
62.41 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11382428
|
Thermo Scientific Alfa Aesar
014481.06 |
5 g |
134.67 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11392428
|
Thermo Scientific Alfa Aesar
014481.14 |
25 g |
536.67 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
88863-33-6 | |
205.80 | |
OHULXNKDWPTSBI-UHFFFAOYSA-N | |
6099237 |
C6H14O2Sr | |
MFCD00050487 | |
strontium isopropoxide, strontium 2+ bis propan-2-olate, strontium propan-2-olate, diisopropoxystrontium, acmc-20egm0, strontium isopropylate, 2-propanol, strontiumsalt 9ci, strontium 2+ ion bis propan-2-olate | |
[Sr++].CC(C)[O-].CC(C)[O-] |
Specifications
88863-33-6 | |
C6H14O2Sr | |
UN3205 | |
strontium isopropoxide, strontium 2+ bis propan-2-olate, strontium propan-2-olate, diisopropoxystrontium, acmc-20egm0, strontium isopropylate, 2-propanol, strontiumsalt 9ci, strontium 2+ ion bis propan-2-olate | |
OHULXNKDWPTSBI-UHFFFAOYSA-N | |
strontium(2+) bis(propan-2-olate) | |
6099237 | |
96% | |
Strontium isopropoxide |
130°C (decomposition) | |
MFCD00050487 | |
Air and moisture sensitive | |
Soluble in isoprpanol. | |
[Sr++].CC(C)[O-].CC(C)[O-] | |
205.80 | |
205.82 | |
Powder |
Safety and Handling
GHS H Statement
H314-H318-H228-H252
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable solid.
Self-heating in large quantities
may catch fire.
P210-P235+P410-P240-P241-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H228-H252-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 4.2; Packing Group: III; Proper Shipping Name: ALKALINE EARTH METAL ALCOHOLATES, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.