missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Strontium Chloride Hexahydrate, puriss. p.a. ACS Reagent, Honeywell Fluka™
Click to view available options
Quantity:
250 g
1 kg
6 x 1 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Quantity | 1 kg |
Linear Formula | SrCl2 · 6H2O |
UN Number | NONH for all modes of transport |
Formula Weight | 266.62g/mol |
Percent Purity | 99 to 102% |
Grade | ACS/puriss. p.a. |
Chemical Name or Material | Strontium Chloride Hexahydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'