missing translation for 'onlineSavingsMsg'
Learn More

Strontium Chloride, 99-102%, Certified AR for Analysis, Fisher Chemical™

49.70 € - 118.00 €

Chemical Identifiers

CAS 10025-70-4
Molecular Formula Cl2H12O6Sr
Molecular Weight (g/mol) 266.61
MDL Number MFCD00149865
InChI Key AMGRXJSJSONEEG-UHFFFAOYSA-L
IUPAC Name strontium(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Sr++]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11423673
SDS View Documents
Fisher Chemical
S/8320/50
250 g N/A N/A N/A
11493723
SDS View Documents
Fisher Chemical
S/8320/53
500 g N/A N/A N/A
Description

Description

Specifications

Chemical Identifiers

10025-70-4
266.61
AMGRXJSJSONEEG-UHFFFAOYSA-L
O.O.O.O.O.O.[Cl-].[Cl-].[Sr++]
Cl2H12O6Sr
MFCD00149865
strontium(2+) hexahydrate dichloride

Specifications

115°C
White
Solid
MFCD00149865
O.O.O.O.O.O.[Cl-].[Cl-].[Sr++]
266.61
Plastic powder jar
10025-70-4
5.0 to 7.0
Cl2H12O6Sr
AMGRXJSJSONEEG-UHFFFAOYSA-L
strontium(2+) hexahydrate dichloride
266.62g/mol
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Strontium chloride hexahydrate

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Strontium Chloride, 99-102%, Certified AR for Analysis, Fisher Chemical™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.