missing translation for 'onlineSavingsMsg'
Learn More

Strontium Carbonate, Extra Pure, SLR, Fisher Chemical™

Product Code. 11418862
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 1633-05-2
Molecular Formula CO3Sr
Molecular Weight (g/mol) 147.63
MDL Number MFCD00011250
InChI Key LEDMRZGFZIAGGB-UHFFFAOYSA-L
Synonym strontium carbonate, strontianite, carbonic acid, strontium salt 1:1, unii-41ypu4mmca, strontium carbonate srco3, ccris 3203, carbonic acid strontium salt 1:1, 41ypu4mmca, strontium, reference standard solution, strontiumcarbonate
PubChem CID 15407
IUPAC Name strontium;carbonate
SMILES [Sr++].[O-]C([O-])=O

Specifications

Color White
Physical Form Solid
Quantity 500 g
Formula Weight 147.6182g/mol
Packaging Plastic powder jar
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'