missing translation for 'onlineSavingsMsg'
Learn More

Gosselin™ Straight Specimen Containers with Blue Cap and Label

CORNING GOSSELIN™ Straight Container, Specimen Vials, PP come in different sizes and quantities, and are available sterile and non-sterile.

Brand:  Gosselin™ TP35C-021

 View more versions of this product

Product Code. 15488814

  • 153.32 € / Pack of 700

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Clear and break-resistant polypropylene (PP)
  • Available in different sizes and quantities
  • Color-coded screw caps
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Specimen Container
Polypropylene
Sterile
Blue
Clear Blue
White label
70/Bag
Yes
Straight
60 mL
Screw Cap
White label
700/Cs.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gosselin™ Straight Specimen Containers with Blue Cap and Label > 60mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.