Learn More
Lenz Laborglasintrumente™ Straight Receiver Adapters
Receiver adapters with drain tip
Brand:  Lenz Laborglasintrumente™ 05272011
Product Code. 11913531
  • 47.00 € / Each
Estimated Shipment: 22-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Receiver adapters acc. to DIN 12594
  • With NS socket and NS cone
  • With drain tip
  • Vacuum connection: thread GL18
  • Made of DURAN™ tubing
Specifications
Specifications
NS 19/26
1
GL 18
Duran Tubing
NS 19/26