missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Stoppers for Serum Bottles and Vials: Snap-on Stopper
Provide airtight covers
316.00 € - 357.00 €
Product Code | Brand | Color | Material | For Use With (Equipment) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Material | For Use With (Equipment) | Price | Quantity & Availability | |||
12492621
|
DWK Life Sciences
224100-080 |
Red | Rubber | Fits 0.7cm I.D., 1.3cm O.D. mouth of serum bottles and vials |
357.00 €
Pack of 1000 |
Estimated Shipment: 25-02-2025
Log in to see stock. |
|||
11571105
|
DWK Life Sciences
W224100-181 |
Gray | Chlorobutyl Rubber | Fits 1.3cm I.D., 2cm O.D. mouth of serum bottles and vials |
316.00 €
Pack of 1000 |
Estimated Shipment: 09-10-2024
Log in to see stock. |
|||
12402641
|
DWK Life Sciences
224100-180 |
Red | Rubber | Fits 1.3cm I.D., 2cm O.D. mouth of serum bottles and vials |
348.00 €
Pack of 1000 |
Estimated Shipment: 29-10-2024
Log in to see stock. |
|||
Description
- Flange-type stoppers are secured with aluminum crimp seals
- Slotted and three-prong plug stoppers are for freeze-dry applications
Specifications
Autoclavable | |
Plug Stopper |