Learn More
Fisherbrand™ Stainless Steel Stirring Shafts with Propellers
Excellent at producing axial flows in vessels from top to bottom. Fisherbrand™ Stainless Steel Stirring Shafts with Propellers are for use with overhead stirrer units.
Brand: Fisherbrand™ 37020-9056
Description
- Available in a range of sizes
- Resistant to corrosion
Specifications
Propellor, 63.5mm | |
1 Each | |
762mm | |
Stirring Shaft |