Learn More
Kimberly-Clark™ Kimtech™ Sterling™ Nitrile-Xtra™ Ambidextrous Gloves
Kimtech™ Sterling™ Nitrile-Xtra™ gloves ensure excellent contamination control with innovative, skin-friendly nitrile suitable for a range of applications. The extra-long gloves are powder- and latex-free, featuring beaded cuffs and textured fingertips.
Specifications
Color Gray
Material Nitrile
View More Specs
Description
Description
  • PPE Cat III according to Regulation (EU) 2016/425
  • EN ISO 374-1:2016 Type C (K) Chemical Splash protection
  • EN 374-4:2003 Resistance to degradation by chemicals
  • EN ISO 374-5:2016 Micro Organism and Virus Protection
  • Manufactured using proprietary Sterling™ technology that combines security and comfort. The efficient, environmentally-friendly construction minimizes waste without compromising safety.
  • Nitrile construction results in products that are stronger and leaner than latex gloves, and feature certified protection against a wide range of contaminants while also being food contact approved.
  • The extra-length gloves are anti-static tested to protect the wearer and equipment, ambidextrous and gray in color.
  • Textured fingertips enhance grip and tactile sensitivity for safer and more efficient processes.
  • Beaded cuffs add strength to the gloves, reducing the risk of tearing and increasing their durability, while also reducing roll down for easier donning and doffing.
  • Contain no natural rubber latex, silicone or powder, reducing the risks of skin irritation for the wearer.
  • Specifications
    Specifications
    Gray
    Nitrile