Learn More
Sampling Systems™ SteriWare Multi-grip Tweezers
SteriWare multi-grip tweezers can be used as normal tweezers, but also have an additional grip feature enabling the easy and secure handling of small vials and containers.
Brand:  Sampling Systems™ 8270A-300
 View more versions of this product
Product Code. 17932062
  • 105.00 € / Pack of 50
Explore more special offers
Description
Description
  • Made from food grade polystyrene
  • Individually bagged to prevent cross contamination
  • 100% manufactured and packed in a cleanroom
  • Available in both sterile and non-sterile
Specifications
Specifications
Serrated, Flat
Polystyrene
Tweezers
Non-sterile
12 in.
303 mm
50 Tweezers/Cs.
Molded