Learn More
Sampling Systems™ SteriWare™ LiquiDip Mono
Sampling Systems™ SteriWare™ LiquiDip Mono is ideal for taking cross sectional or point samples of liquids.
Brand:  Sampling Systems™ 8920H-1100
Product Code. 15859571
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Assembled and packed in a Class 100,000 Cleanroom
  • Individually bagged
  • Made from FDA, EC 1935/2004 and EU 10/2011 conforming HDPE
  • BSE/TSE free
  • Suitable for taking cross sectional samples
  • Full batch traceability
  • Available pre-sterilized (SAL 10-6)
  • Available in 2 different lengths
  • Push the sampler into the product, ensuring the sampler is in the open position
  • Push the sampler against the base of the container to close the valve
  • Remove the sampler and pour out the liquid into a suitable container
  • No cross contamination
  • No cleaning required
Specifications
Specifications
Liquid Sampler
Valve at one end
Single-use
21 mm
HDPE
225 mL
Non-sterile
1100 mm