Learn More
Sampling Systems™ SteriWare™ Funnels
BIOPHARMA
SteriWare™ Funnels are part of a large range of high quality single use liquid funnels. A large range of sizes in sterile and non sterile mean that we will have the right item for you. Simply unwrap and use.
Brand:  Sampling Systems™ 8370H-50
Product Code. 17594656
  • 115.00 € / Pack of 100
Estimated Shipment: 14-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Disposable, single use, sterile liquid funnels.
  • They are ideal for use in laboratories and in manufacturing areas.
  • These funnels are made from food grade polymers and are designed to be used straight from the bag.
  • SteriWare™ Funnels are available sterile and non-sterile.
  • Simply unwrap and use.
  • They are ideal for dispensing liquids and creams.
  • Each funnel has a small lug on the rim so you can hold the funnel and not get your fingers wet.
  • Made from high quality HDPE which conforms to the latest FDA and EU food requirements.
  • BSE free.
  • Cleanroom manufactured.
  • Available sterile (SAL 10-6) and non-sterile.
  • 50 mm, 100 mm and 200 mm top diameters.
  • All orders are supplied with certificates of conformity and sterilisation where applicable.
Specifications
Specifications
50 mm
HDPE
Non-sterile