missing translation for 'onlineSavingsMsg'
Learn More

Sampling Systems™ SteriWare™ Disposable ViscoThief
BIOPHARMA

Sampling Systems™ SteriWare™ Disposable ViscoThief is a single use, disposable sampler that is designed for use with viscous materials such as creams, oils, and honey.

Brand:  Sampling Systems™ 8055H-1500

Product Code. 15809451

  • 282.00 € / Pack of 20
Estimated Shipment: 13-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Assembled and packed in a Class 100,000 Cleanroom
  • Individually bagged
  • Made from FDA, EC 1935/2004 and EU 10/2011 conforming materials
  • BSE/TSE free
  • Full batch traceability
  • Available pre-sterilized (SAL 10-6)
  • At the required depth pull up the handle
  • The further the handle is pulled, the larger the sample volume
  • Push handle down to expel the sample
  • No cross contamination
  • No cleaning required
Specifications

Specifications

Liquid Sampler
Individually bagged
Single-use
21 mm
HDPE
300 mL
Non-sterile
1500 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.