Learn More
Sampling Systems™ SteriWare™ Disposable Tray
BIOPHARMA
Sampling Systems™ SteriWare™ Disposable Tray is ideal for hygienic applications within hospitals and pharmaceutical factories.
Brand:  Sampling Systems™ 8081P-3000
 View more versions of this product
Product Code. 15819461
  • 265.00 € / Pack of 40
Estimated Shipment: 24-10-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Moulded and packed in a Class 100,000 Cleanroom
  • Individually bagged
  • Made from FDA, EC 1935/2004 and EU 10/2011 conforming material
  • BSE/TSE free
  • Full batch traceability
  • Available pre-sterilized (SAL 10-6)
  • No cross contamination
  • No cleaning required
Specifications
Specifications
Polypropylene
Single-use
White
3 L
Non-sterile
310 x 265 x 55 mm