Learn More
Sampling Systems™ SteriWare™ Disposable Powder Probe
BIOPHARMA
Sampling Systems™ SteriWare™ Disposable Powder Probe is ideal for taking a cross sectional sample through a bulk powder.
Brand:  Sampling Systems™ 8064H-500
 View more versions of this product
Product Code. 15849451
  • 108.00 € / Pack of 20
Estimated Shipment: 10-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Assembled and packed in a Class 100,000 Cleanroom
  • Individually bagged
  • Made from FDA, EC 1935/2004 and EU 10/2011 conforming HDPE
  • BSE/TSE free
  • Full batch traceability
  • Available pre-sterilized (SAL 10-6)
  • Suitable for use in ATEX areas
  • Push the sampler into the product, rotate the sampler then withdraw
  • Deposit the sample into a suitable container
  • Place the used sampler into its original plastic bag and dispose
  • No cross contaminatin
  • No cleaning required
Specifications
Specifications
Probe
500mm Blade
125 mL
Powder Sampling
HDPE
Prevents sample from falling out
Non-sterile
600 mm