Learn More
Sampling Systems™ SteriWare™ Disposable Jumbo Pipette
Sampling Systems™ SteriWare™ Disposable Jumbo Pipette is both quick and easy to use and is ideal for taking samples from low viscosity liquids.
Brand:  Sampling Systems™ 8051H-1100S
Product Code. 15859441
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Assembled and packed in a Class 100,000 Cleanroom
  • Individually bagged
  • Made from FDA, EC 1935/2004 & EU 10/2011 conforming materials
  • BSE/TSE free
  • Full batch traceability
  • Available pre-sterilized (SAL 10-6)
  • Highly visible color coded handle
  • Press thumb onto blue end of the tube until lowered to the correct sampling depth
  • At the required depth raise thumb until levels equalize
  • Press the thumb onto tube to seal in the sample
  • No cross contamination
  • No cleaning required
Specifications
Specifications
HDPE
225 mL
21 mm
Individually Bagged
Sterile
1100 mm