missing translation for 'onlineSavingsMsg'
Learn More

Sampling Systems™ SteriWare™ Counting Triangle
BIOPHARMA

Sampling Systems™ SteriWare™ Counting Triangle is ideal for counting round tablets.

Brand:  Sampling Systems™ 8066A-1

Product Code. 15890098

  • 114.00 € / Pack of 30
Estimated Shipment: 10-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Moulded and packed in a Class 100,000 Cleanroom
  • Individually bagged
  • Made from FDA, EC 1935/2004 and EU 10/2011 conforming HDPE
  • BSE/TSE free
  • Full batch traceability
  • No cross contamination
  • No cleaning required
Specifications

Specifications

Non-sterile
230 mm
Counting Triangle
30
13 mm
Polystyrene
230 mm
Triangular
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.