missing translation for 'onlineSavingsMsg'
Learn More

SP Bel-Art™ Sterileware Upright Handle Dippers/Ladles

Available in blue or white to allow for effective sample contrast, these sterile ladles are useful for sampling liquids and creams

Brand:  SP Bel-Art™ F36775-1050

 View more versions of this product

Product Code. 16886896

  • 143.00 € / Pack of 40
Estimated Shipment: 18-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Upright handle allows for sampling from above container
  • Curved handle end provides secure withdrawal of sample
  • Fits into 2 in. openings such as a 45-gallon drum bunghole
  • Made from FDA and EU conforming materials; safe for food, drug and cosmetic use
  • White high density polyethylene (HDPE), or blue polypropylene (PP) ladles heat sealed in individual polyethylene bags
  • Gamma irradiated sterile (SAL 10^-6)
  • Lot stamped for accurate tracing; shipped with a Certificate of Processing for Sterilization
Specifications

Specifications

50 mL
White
40/Pk.
26 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers