Learn More
Statsafe^t 6000, Thermo Scientific Chemicals
Specifications
Physical Form | Liquid |
---|---|
Boiling Point | 94°C |
Flash Point | <−5°C (−23°F) |
UN Number | UN1993 |
Solubility Information | Not miscible with water. |
Description
Static dissipater used in many chemical processing operations to reduce the risk of static build up and consequent damage and loss. Especially recommended for use in the manufacturing of polyolefins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsStatic dissipater used in many chemical processing operations to reduce the risk of static build up and consequent damage and loss. The Protein Crystallography Extension Kit substantially increases the screening range of the Basic Crystallography Kit by providing a broader range of organic solvents, PEGs, and additives (divalent ions and stabilizers).
Solubility
Not miscible with water.
Notes
Suitable additive to increase hydrocarbon conductivity and to reduce electrostatic hazards in a variety of applications.
Specifications
Liquid | |
<−5°C (−23°F) | |
Not miscible with water. |
94°C | |
UN1993 |
Safety and Handling
GHS H Statement
H225-H304-H314-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c
H225-H304-H315-H319-H336
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only