Learn More
IBA Lifesciences StarGate pPSG-IBA Acceptor Vectors for E. coli Expression
Achieves high-level expression in E. coli by the T7 promoter by providing specified genetic surroundings for optimal expression and purification of non-toxic proteins.
220.00 €
Specifications
Format | Liquid |
---|---|
For Use With (Equipment) | StarGate™ System, E. Coli BL21(DE3) |
Inducing Agent | E. coli host with T7 RNA polymerase gene (e.g. BL21(DE3)) |
pH | 8 |
Promoter | T7 |
Product Code | Brand | Description | Protein Tag | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Protein Tag | Price | Quantity & Availability | ||||
15633758
|
IBA Lifesciences
5-4200-001 |
pPSG-IBA wt1 | Untagged | N/A | |||||
15643758
|
IBA Lifesciences
5-4203-001 |
pPSG-IBA 3 | Strep-tag for C-terminal | N/A | |||||
15653758
|
IBA Lifesciences
5-4205-001 |
pPSG-IBA 5 | Strep-tag for N-terminal | N/A | |||||
15663758
|
IBA Lifesciences
5-4223-001 |
pPSG-IBA 23 | Strep-tag for C-terminal and GST-tag for N-terminal | N/A | |||||
15673758
|
IBA Lifesciences
5-4225-001 |
pPSG-IBA 25 | GST Tag for N-terminal | N/A | |||||
15683758
|
IBA Lifesciences
5-4233-001 |
pPSG-IBA 33 | His Tag for C-terminal | N/A | |||||
15693758
|
IBA Lifesciences
5-4235-001 |
pPSG-IBA 35 | His Tag (6x) for N-terminal | N/A | |||||
15603768
|
IBA Lifesciences
5-4243-001 |
pPSG-IBA 43 | Strep-tag for C-terminal and His Tag (6x) for N-terminal | N/A | |||||
15613768
|
IBA Lifesciences
5-4245-001 |
pPSG-IBA 45 | Strep-tag for N-terminal and His Tag (6x) for C-terminal | N/A | |||||
15623768
|
IBA Lifesciences
5-4262-001 |
pPSG-IBA 62 | Strep-tag for C-terminal and FLAG-tag for N-terminal | N/A | |||||
15633768
|
IBA Lifesciences
5-4263-001 |
pPSG-IBA 63 | FLAG-tag for C-terminal | N/A | |||||
15643768
|
IBA Lifesciences
5-4264-001 |
pPSG-IBA 64 | Strep-tag for N-terminal and FLAG-tag for C-terminal | N/A | |||||
15653768
|
IBA Lifesciences
5-4265-001 |
pPSG-IBA 65 | FLAG-tag for N-terminal | N/A | |||||
15663768
|
IBA Lifesciences
5-4303-001 |
pPSG-IBA 103 | Twin-Strep-tag for C-terminal | N/A | |||||
15673768
|
IBA Lifesciences
5-4305-001 |
pPSG-IBA 105 | Twin-Strep-tag for N-terminal | N/A | |||||
15683768
|
IBA Lifesciences
5-4323-001 |
pPSG-IBA 123 | Twin-Strep-tag for C-terminal and GST-tag for N-terminal | N/A | |||||
15693768
|
IBA Lifesciences
5-4343-001 |
pPSG-IBA 143 | Twin-Strep-tag for C-terminal and His Tag (6x) for N-terminal | N/A | |||||
15603778
|
IBA Lifesciences
5-4345-001 |
pPSG-IBA 145 | Twin-Strep-tag for N-terminal and His Tag (6x) for C-terminal | N/A | |||||
15613778
|
IBA Lifesciences
5-4362-001 |
pPSG-IBA 162 | Twin-Strep-tag for C-terminal and FLAG-tag for N-terminal | N/A | |||||
15623778
|
IBA Lifesciences
5-4364-001 |
pPSG-IBA 164 | Twin-Strep-tag for N-terminal and FLAG-tag for C-terminal | N/A | |||||
15633778
|
IBA Lifesciences
5-4367-001 |
pPSG-IBA 167 | Twin-Strep-tag/FLAG-tag for N-terminal | N/A | |||||
15643778
|
IBA Lifesciences
5-4368-001 |
pPSG-IBA 168 | Twin-Strep-tag/FLAG-tag for C-terminal | N/A | |||||
Description
pPSG-IGA E. coli expression vectors:
pPSG-IBA vectors use the T7 promoter and T7 RNA polymerase for high-level transcription of the gene of interest.
- High-level expression in E. coli by bacteriophage T7 promoter
- High-level transcription by T7 RNA polymerase in BL21 strains
- High-level expression of non-toxic proteins
- Induction by IPTG
- Suitable for in vitro transcription/translation
StarGate System
IBA developed StarGate to provide a simple, fast and highly efficient cloning process for genes of interest (GOI) for use in functional expression, purification, detection, immobilization or separation of recombinant proteins:
- Minimal extra modification of the gene due to short combinatorial sites
- Inherent high-level cloning efficiency due to a directed reaction (no equilibrium)
- GOIs are cloned to obtain a Donor Vector; 2 or more genes can be combined using Fusion Cloning
- Genes are then transferred into Acceptor Vectors that provide optimal genetic surroundings
- The final expression vector, the Destination Vector, is then placed into the respective host
Specifications
Liquid | |
E. coli host with T7 RNA polymerase gene (e.g. BL21(DE3)) | |
T7 | |
20 μL TE buffer: 10 mm Tris-HCl, 1 mm EDTA / 5 μg | |
250 ng/μL | |
Total length: 3469 | |
Cloning genes of interest for use in functional expression, purification, detection, immobilization or separation of recombinant proteins |
StarGate™ System, E. Coli BL21(DE3) | |
8 | |
Acceptor Vector for E. coli expression | |
2°C to 8°C for frequent usage, -20°C for long-term storage | |
5 μg | |
Clear |
Safety and Handling
missing translation for 'shelfLife' : 12 months
Your input is important to us. Please complete this form to provide feedback related to the content on this product.