missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Standard Buffer Solution pH 1.68, SI Analytics™
Click to view available options
Quantity:
250 mL
60 Ampules
Description
- Includes manufacturer certificate
- Mixture, consisting of the following components: Water, potassium tetraoxalate dihydrate
- Store tigthly sealed at temperatures between 15°C and 25°C
- The product does not contain any relevant quantities of materials with critical values that have to be monitored at the workplace
- Rinse off of bigger amounts into drains or the aquatic environment may lead to decreased pH-values. A low pH-value harms aquatic organisms. In the dilution of the use-level the pH-value is considerably increased, so that after the use of the product the aqueous waste, emptied into drains, is only low water-dangerous
Specifications
Specifications
Boiling Point | 100°C |
Color | Colorless |
Density | 1.01g/cm3 |
pH | 1.68 |
Physical Form | Liquid |
Quantity | 60 Ampules |
Formulation | Water, potassium tetraoxalate dihydrate |
Odor | Odorless |
Packaging | FIOLAX™ Ampules |
Sterility | Sterile |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction