Learn More
Thermo Scientific Chemicals Stains-all, 97%, pure
40.90 € - 326.63 €
Chemical Identifiers
CAS | 7423-31-6 |
---|---|
Molecular Formula | C30H28BrN2S2+ |
Molecular Weight (g/mol) | 560.592 |
MDL Number | MFCD00078788 |
InChI Key | MPBRYMWMMKKRGC-UHFFFAOYSA-N |
Synonym | 1-Ethyl-2-[3-(1-ethylnaphtho[1, 2-d]thiazolin-2-ylidene)-2-methylpropenyl]naphtho[1, 2-d]thiazolium bromide |
PubChem CID | 123134119 |
IUPAC Name | (2E)-1-ethyl-2-[(E)-3-(1-ethylbenzo[e][1,3]benzothiazol-1-ium-2-yl)-2-methylprop-2-enylidene]benzo[e][1,3]benzothiazole;hydrobromideShow More |
SMILES | CCN1C(=CC(=CC2=[N+](C3=C(S2)C=CC4=CC=CC=C43)CC)C)SC5=C1C6=CC=CC=C6C=C5.Br |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10370594
|
Thermo Scientific Chemicals
213512500 |
250 mg | Glass bottle |
40.90 €
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10276840
|
Thermo Scientific Chemicals
213510010 |
1 g | Glass bottle |
98.39 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10410491
|
Thermo Scientific Chemicals
213510050 |
5 g | Glass bottle |
326.63 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
Specifications
7423-31-6 | |
C30H28BrN2S2+ | |
27, IV, 8619 | |
MPBRYMWMMKKRGC-UHFFFAOYSA-N | |
CCN1C(=CC(=CC2=[N+](C3=C(S2)C=CC4=CC=CC=C43)CC)C)SC5=C1C6=CC=CC=C6C=C5.Br | |
560.592 | |
559.59 | |
Pure | |
Gray to Purple | |
Stains-all, 97% |
97% | |
MFCD00078788 | |
1-Ethyl-2-[3-(1-ethylnaphtho[1, 2-d]thiazolin-2-ylidene)-2-methylpropenyl]naphtho[1, 2-d]thiazolium bromide | |
Authentic | |
(2E)-1-ethyl-2-[(E)-3-(1-ethylbenzo[e][1,3]benzothiazol-1-ium-2-yl)-2-methylprop-2-enylidene]benzo[e][1,3]benzothiazole;hydrobromide | |
123134119 | |
97% | |
Complies | |
Powder |
Safety and Handling
LIGHT SENSITIVE
missing translation for 'einecsNumber' : 231-047-7
missing translation for 'tsca' : TSCA
- Stains-all
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.