Learn More
RETSCH Stainless-Steel Test Sieves, 200 Dia. x 50 mmH, with Perforated Plates with Square Holes
Brand:  RETSCH 60.190.056000
 View more versions of this product
Product Code. 10778624
  • 214.00 € / Each
Estimated Shipment: 28-02-2025
to see stock.
Explore more special offers
Add to basket
Specifications
1 Each
Test Sieve
Certificate indicating Compliance
For separation, fractioning, particle size determination
Stainless Steel
56 mm
ISO 3310/2
50mm
200 mm
Stainless Steel