Learn More
RETSCH Stainless-Steel Test Sieves, 200 Dia. x 50 mmH, with Perforated Plates with Square Holes
Brand:  RETSCH 60.190.005000
 View more versions of this product
Product Code. 10193812
  • 221.00 € / Each
Estimated Shipment: 21-03-2025
to see stock.
Explore more special offers
Add to basket
Specifications
1 Each
Test Sieve
Certificate indicating Compliance
For separation, fractioning, particle size determination
Stainless Steel
5 mm
ISO 3310/2
50mm
200 mm
Stainless Steel