Learn More
Bel-Art™ SP Scienceware™ Stainless-steel Spoons
Double-ended with two bowl sizes.
Brand:  Bel-Art™ F36730-0021
 View more versions of this product
Product Code. 11716545
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Double-ended with two bowl sizes.
  • Made from 304 stainless-steel
  • Sampling spoons have solid, rounded shaft
  • Available in plain polished stainless-steel or coated with Fluo-Kem™ PTFE FEP
Specifications
Specifications
Spoon
1.6, 3 mL
Brown
21 cm
Stainless Steel
Solid, rounded shaft
Fluo-Kem Teflon FEP coated